missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Earth's Processes Poster

Visualize key geologic processes.

Manufacturer:  Fisher Scientific AZ351035

 View more versions of this product

Catalog No. S65139



Description

Description

  • Detailed overview of the rock cycle as well as key Earth processes
  • Faulting and folding, earthquakes, volcanism, weathering, erosion, and more
  • Dimensions: 23W × 35 in.H (58.4 × 88.9cm)

Specifications

Specifications

Geology
35 in.
Paper
23 in.
Detailed overview of the rock cycle as well as such key earth processes as faulting and folding, earthquakes, volcanism, weathering, erosion and more
88.9cm
Earth's Processes Poster
58.42cm
Documents