Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution, without sodium bicarbonate


Quantity 100mL

Manufacturer: alfa aesar™  J67590AE

 View more versions of this product

Catalog No. AAJ67590AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Earle's Balanced Salts Solution
Color Colorless
Quantity 100mL
TSCA TSCA
Name Note without sodium bicarbonate
Physical Form Liquid
MDL Number MFCD00217343
Synonym EBSS
Odor Odorless

Specifications

Chemical Name or Material Earle's Balanced Salts Solution
Name Note without sodium bicarbonate
MDL Number MFCD00217343
Synonym EBSS