Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution, without calcium chloride, without magnesium sulfate

Quantity 100mL

Manufacturer: alfa aesar™  J67539-AE

 View all options for this product

Catalog No. AAJ67539AE

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications

Chemical Identifiers

Odor Odorless
Quantity 100mL
Color Colorless
Quantity 100mL
Chemical Name or Material Earle's Balanced Salts Solution
Name Note Without calcium chloride, without magnesium sulfate
Physical Form Liquid
MDL Number MFCD00217343
Synonym EBSS