missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution

Specifications

Chemical Name or Material Earle'sBalancedSaltsSolution
MDL Number MFCD00217343
Synonym EBSS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67618K2
View Documents
Alfa Aesar™
J67618K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67618AP
View Documents
Alfa Aesar™
J67618AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Earle'sBalancedSaltsSolution
MFCD00217343
EBSS