Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution


Quantity 100mL

Manufacturer: alfa aesar™  J67618AE

 View more versions of this product

Catalog No. AAJ67618AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Earle's Balanced Salts Solution
MDL Number MFCD00217343
Synonym EBSS
Physical Form Liquid
Quantity 100mL
Odor Odorless
Color Colorless
TSCA TSCA

Specifications

Chemical Name or Material Earle's Balanced Salts Solution
MDL Number MFCD00217343
Synonym EBSS