missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X), without sodium bicarbonate

Specifications

Chemical Name or Material Earle'sBalancedSaltsSolution(10X)
Name Note without sodium bicarbonate
MDL Number MFCD00217343
Synonym EBSS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67697AP
View Documents
Alfa Aesar™
J67697AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67697K2
View Documents
Alfa Aesar™
J67697K2
1L N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Specifications

Earle'sBalancedSaltsSolution(10X)
without sodium bicarbonate
MFCD00217343
EBSS