Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X), without sodium bicarbonate


Quantity 500mL

Manufacturer: alfa aesar™  J67697AP

 View more versions of this product

Catalog No. AAJ67697AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
MDL Number MFCD00217343
Name Note without sodium bicarbonate
Synonym EBSS
TSCA TSCA
Quantity 500mL
Odor Odorless
Color Colorless
Chemical Name or Material Earle's Balanced Salts Solution (10X)

Specifications

Chemical Name or Material Earle's Balanced Salts Solution (10X)
Name Note without sodium bicarbonate
MDL Number MFCD00217343
Synonym EBSS