missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X), without magnesium sulfate

Chemical Identifiers

MDL Number MFCD00217343
Synonym EBSS
Chemical Name or Material Earle's Balanced Salts Solution (10X), without magnesium sulfate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67567AE SDS Alfa Aesar™
J67567AE
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67567AP SDS Alfa Aesar™
J67567AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67567K2 SDS Alfa Aesar™
J67567K2
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00217343
EBSS
Earle's Balanced Salts Solution (10X), without magnesium sulfate

Specifications

Liquid
EBSS
Colorless
Earle's Balanced Salts Solution (10X), without magnesium sulfate
MFCD00217343
Odorless
100mL

Safety and Handling

TSCA : TSCA