Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X), without calcium chloride


Quantity 100mL

Manufacturer: alfa aesar™  J67782-AE

 View more versions of this product

Catalog No. AAJ67782AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Odor Odorless
Color Colorless
Quantity 100mL
Chemical Name or Material Earle's Balanced Salts Solution (10X)
Name Note without calcium chloride
Physical Form Liquid
MDL Number MFCD00217343
Synonym EBSS
TSCA TSCA

Specifications

Chemical Name or Material Earle's Balanced Salts Solution (10X)
Name Note without calcium chloride
MDL Number MFCD00217343
Synonym EBSS