missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X), without calcium chloride

Chemical Identifiers

MDL Number MFCD00217343
Synonym EBSS
Chemical Name or Material Earle's Balanced Salts Solution (10X), without calcium chloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67782AP SDS Alfa Aesar™
J67782AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67782K2 SDS Alfa Aesar™
J67782K2
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00217343
EBSS
Earle's Balanced Salts Solution (10X), without calcium chloride

Specifications

Liquid
EBSS
Colorless
Earle's Balanced Salts Solution (10X), without calcium chloride
MFCD00217343
Odorless
500mL