Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X)

Chemical Identifiers

Chemical Name or Material Earle's Balanced Salts Solution (10X)
MDL Number MFCD00217343
Synonym EBSS
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67634AP

  SDS
alfa aesar™
J67634AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67634K2

  SDS
alfa aesar™
J67634K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Earle's Balanced Salts Solution (10X)
MDL Number MFCD00217343
Physical Form Liquid
TSCA TSCA
Quantity 500mL
Odor Odorless
Color Colorless
Synonym EBSS