Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X)


Quantity 1L

Manufacturer: alfa aesar™  J67634K2

 View more versions of this product

Catalog No. AAJ67634K2

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Earle's Balanced Salts Solution (10X)
MDL Number MFCD00217343
Synonym EBSS
Color Colorless
Quantity 1L
TSCA TSCA
Physical Form Liquid
Odor Odorless

Specifications

Chemical Name or Material Earle's Balanced Salts Solution (10X)
MDL Number MFCD00217343
Synonym EBSS