Please login to your online account to display your discounted pricing

Alfa Aesar™ Earle's Balanced Salts Solution (10X)


Quantity 500mL

Manufacturer: alfa aesar™  J67634AP

 View more versions of this product

Catalog No. AAJ67634AP

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Earle's Balanced Salts Solution (10X)
MDL Number MFCD00217343
Physical Form Liquid
TSCA TSCA
Quantity 500mL
Odor Odorless
Color Colorless
Synonym EBSS

Specifications

Chemical Name or Material Earle's Balanced Salts Solution (10X)
MDL Number MFCD00217343
Synonym EBSS