Please login to your online account to display your discounted pricing
Eagle™ Thermoplastics™ Safety Bottle Carriers
Safety bottle carriers are molded-in handle
$217.50 - $225.68
Specifications
Autoclavable No
Material Plastic
View More Specs
Products
Catalog Number Mfr. No. Color Price Quantity    
03-439A
fisher scientific
B100
Red Each for $225.68
03-439
fisher scientific
B102
Black Each for $217.50
Description & Specifications
Specifications
Autoclavable No
Material Plastic
  • Thick molded rubber safely cushions glass bottles
  • Circular cup in the bottom centers bottle with 1 in. (2.5cm) between wall and bottle
  • Integral molded handle