Please login to your online account to display your discounted pricing

Eact, Tocris™

Activator of Ca2+-activated Cl- channel transmembrane protein 16A (TMEM16A)

$218.74 - $911.19

Chemical Identifiers

Chemical Name or Material Eact
Assay Percent Range >99%
Molecular Formula C22H24N2O5S
Formula Weight 428.5
Synonym 3, 4, 5-Trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-2-thiazolyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

487650

 
r & d systems
RDS487650
50mg Each for $911.19

487610

 
r & d systems
RDS487610
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 428.5
Quantity 50mg
Chemical Name or Material Eact
Assay Percent Range >99%
Molecular Formula C22H24N2O5S
CAS 461000-66-8
Recommended Storage Store at +4°C
Synonym 3, 4, 5-Trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-2-thiazolyl)benzamide
Safety and Handling

Store at +4°C