Please login to your online account to display your discounted pricing

E4CPG, Tocris

Group I/group II mGlu antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material E4CPG
Assay Percent Range >99%
Molecular Formula C11H13NO4
Formula Weight 223.23
Synonym (RS)-α-Ethyl-4-carboxyphenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

100950

 
r & d systems
RDS100950
50mg Each for $746.48

100910

 
r & d systems
RDS100910
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 223.23
Quantity 50mg
CAS 170846-89-6
Chemical Name or Material E4CPG
Molecular Formula C11H13NO4
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Synonym (RS)-α-Ethyl-4-carboxyphenylglycine
Safety and Handling

Store at Room Temperature