Please login to your online account to display your discounted pricing

E4CPG, Tocris


Group I/group II mGlu antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS100950

 View more versions of this product

Catalog No. 100950

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 223.23
Quantity 50mg
CAS 170846-89-6
Chemical Name or Material E4CPG
Molecular Formula C11H13NO4
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Synonym (RS)-α-Ethyl-4-carboxyphenylglycine

Specifications

Chemical Name or Material E4CPG
Assay Percent Range >99%
Molecular Formula C11H13NO4
Formula Weight 223.23
Synonym (RS)-α-Ethyl-4-carboxyphenylglycine
Accessories

Store at Room Temperature