Please login to your online account to display your discounted pricing

E4BP4 (NFIL3) Rat anti-Mouse, PE, Clone: S2M-E19, eBioscience™

Rat Monoclonal Antibody

$172.55 - $448.05

Specifications

Antigen E4BP4 (NFIL3)
Clone S2M-E19
Host Species Rat
Gene Alias NFIL3A
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122352

 
life technologies
LS12592780
25μg Each for $172.55

501122353

 
life technologies
LS12592782
100μg Each for $448.05
Description & Specifications

Specifications

Antigen E4BP4 (NFIL3)
Clone S2M-E19
Host Species Rat
Gene Alias NFIL3A
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This S2M-E19 monoclonal antibody recognizes mouse E4BP4 (NFIL3), a basic leucine zipper transcription factor first identified for its binding to the promoters of the adenovirus E4 protein and human IL-3. E4BP4 is involved in the transcriptional control of the mammalian circadian clock, motor neuron growth and survival, regulation of von Willbrand factor expression, and osteoblast function. In addition, it is also required for the development of NK and NKT cells as well as CD8a+ dendritic cells. Although relatively high levels of mRNA are reported in resting NK and NKT cells, studies at eBioscience have observed expression of E4BP4 protein in lymphocytes only after activation.