Please login to your online account to display your discounted pricing

E4BP4 (NFIL3) Mouse anti-Human, PE, Clone: MABA223, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $448.05

Specifications

Antigen E4BP4 (NFIL3)
Clone MABA223
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122461

 
life technologies
LS12981241
25 tests Each for $172.55

501122462

 
life technologies
LS12981242
100 tests Each for $448.05
Description & Specifications

Specifications

Antigen E4BP4 (NFIL3)
Clone MABA223
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This MABA223 monoclonal antibody recognizes human E4BP4 (NFIL3), a basic leucine zipper transcription factor first identified for its binding to the promoters of the adenovirus E4 protein and human IL-3. E4BP4 is involved in the transcriptional control of the mammalian circadian clock, motor neuron growth and survival, regulation of von Willbrand factor expression, and osteoblast function. In addition, it is required for the development of NK and NKT cells as well as CD8a+ dendritic cells. Although relatively high levels of mRNA are reported in resting NK and NKT cells, studies at eBioscience have observed expression of E4BP4 protein in lymphocytes only after activation.