Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ E2F-4 Transcription Factor Ab-4, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and gel supershift experiments.

Manufacturer: thermo scientific™ lab vision™  MS1057PABX

 View more versions of this product

Catalog No. MS1057PABX

  • / Each
 

Description & Specifications

Specifications

Format Concentrated
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Rat
Immunogen Recombinant human E2F-4 protein
Antigen E2F-4 Transcription Factor Ab-4
Host Species Mouse
Quantity 200µL
Clone 4E2F04; same as WUF10
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Transcription Regulation
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-4 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-4 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides. Differential phosphorylation contributes to the microheterogeniety of E2F-4 (total aa 411-416, varies due to a trinucleotide repeat).

Host Species: Mouse

Clone: 4E2F04; Same as WUF10

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant human E2F-4 protein

Molecular Weight: ∽56 kDa

Positive Control: Raji cells, Tonsil

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO