Please login to your online account to display your discounted pricing

Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

$318.84 - $1,565.71

Specifications

Antigen E2F-3 Transcription Factor Ab-4
Clone 3E2F04; same as PF37
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1063P0

 
thermo scientific™
MS1063P0
200μg/mL 100µL Each for $318.84

MS1063P1

 
thermo scientific™
MS1063P1
200μg/mL 500µL Each for $875.61

MS1063P1ABX

 
thermo scientific™
MS1063P1ABX
1mg/mL 100µL Each for $1,000.57

MS1063PABX

 
thermo scientific™
MS1063PABX
1mg/mL 200µL Each for $1,565.71
Description & Specifications

Specifications

Antigen E2F-3 Transcription Factor Ab-4
Clone 3E2F04; same as PF37
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Transcription Regulation
Type Primary
Immunogen Recombinant human E2F-3 protein
Isotype IgG2a

Obtain accurate, reproducible results in immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-3 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides

Host Species: Mouse

Clone: 3E2F04; same as PF37

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human E2F-3 protein

Molecular Weight: ∽58 kDa

Positive Control: Raji cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO