Please login to your online account to display your discounted pricing

Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS1063PABX

 View more versions of this product

Catalog No. MS1063PABX

  • / Each
 

Description & Specifications

Specifications

Antigen E2F-3 Transcription Factor Ab-4
Applications Immunofluorescence
Classification Monoclonal
Clone 3E2F04; same as PF37
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Recombinant human E2F-3 protein
Isotype IgG2a
Quantity 200μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Obtain accurate, reproducible results in immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-3 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides

Host Species: Mouse

Clone: 3E2F04; same as PF37

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human E2F-3 protein

Molecular Weight: ∽58 kDa

Positive Control: Raji cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO