Please login to your online account to display your discounted pricing

Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™  MS1063P1ABX

 View more versions of this product

Catalog No. MS1063P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen E2F-3 Transcription Factor Ab-4
Format Concentrated
Applications Immunofluorescence
Isotype IgG2a
Quantity 100µL
Host Species Mouse
Immunogen Recombinant human E2F-3 protein
Regulatory Status RUO
Clone 3E2F04; same as PF37
Research Discipline Transcription Regulation
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Obtain accurate, reproducible results in immunofluorescence and gel supershift experiments with Thermo Scientific™ E2F-3 Transcription Factor Ab-4, Mouse Monoclonal Antibody.

E2F transcription factors are functionally regulated by binding to Rb p110, p107, and p130. E2F-3 is regulated by complex formation with Rb p110. E2F family members bind DNA as heterodimers with members of the DP family of polypeptides

Host Species: Mouse

Clone: 3E2F04; same as PF37

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human E2F-3 protein

Molecular Weight: ∽58 kDa

Positive Control: Raji cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Immunofluorescence
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO