Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2% DFS Item
System starter for fast and convenient E-Gel agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel SYBR Safe 1.2% agarose gels.
Manufacturer:  Invitrogen™ LSG8351ST
Catalog No. G8351ST
Includes: E-Gel Power Snap Electrophoresis device,E-Gel Power Snap Camera,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
Description
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, SYBR Safe 1.2%, provides everything you need to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
  • 18 E-Gel agarose gels with SYBR Safe, 1.2% (Cat. No. G521801)
  • E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)
Warranty
1 year warranty
Specifications
Specifications
240 V ac (Power Supply), 48 V dc (Electrophoresis Device)
E-Gel™
Electrophoresis System Starter Kit
Room Temperature
SDS
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.