Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2% DFS Item
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel SYBR Safe 2% agarose gels.
Manufacturer:  Invitrogen™ LSG8152ST
Catalog No. G8152ST
Includes: E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2%, provides everything you need to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the device.
The kit includes:
  • 18 E-Gel agarose gels with SYBR Safe, 2% (Cat. No. G521802)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
Warranty
1 year warranty
Specifications
Specifications
Mini
E-Gel™
Room Temperature
240 V ac (Power Supply), 48 V dc (Electrophoresis Device)
Electrophoresis Device Starter Kit
SDS
Documents
Product Certifications
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.