missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (E)-alpha-Methylstilbene, 98%

Manufacturer:  Alfa Aesar™ A1535122

 View more versions of this product

Catalog No. AAA1535122



Specifications

Specifications

(E)-alpha-Methylstilbene
100g
C15H14
1906425
OVZXISBUYCEVEV-OUKQBFOZSA-N
[(E)-1-phenylprop-1-en-2-yl]benzene
1549166
98%
79°C to 82°C
833-81-8
MFCD00026343
1,2-diphenylpropene, e-alpha-methylstilbene, .alpha.-methylstilbene, 1,2-diphenyl-1-propene, 1-methyl-1,2-diphenylethene, stilbene, .alpha.-methyl, 1-propene, 1,2-diphenyl, 1-methyl-2-phenylvinyl benzene, trans-alpha-methylstilbene, chembl14773
CC(=CC1=CC=CC=C1)C2=CC=CC=C2
194.277
194.28

Chemical Identifiers

833-81-8
194.277
OVZXISBUYCEVEV-OUKQBFOZSA-N
1549166
CC(=CC1=CC=CC=C1)C2=CC=CC=C2
C15H14
MFCD00026343
1,2-diphenylpropene, e-alpha-methylstilbene, .alpha.-methylstilbene, 1,2-diphenyl-1-propene, 1-methyl-1,2-diphenylethene, stilbene, .alpha.-methyl, 1-propene, 1,2-diphenyl, 1-methyl-2-phenylvinyl benzene, trans-alpha-methylstilbene, chembl14773
[(E)-1-phenylprop-1-en-2-yl]benzene
Documents