Please login to your online account to display your discounted pricing

Alfa Aesar™ (E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester, 98%

$113.28 - $403.30

Chemical Identifiers

Chemical Name or Material (E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester
CAS 173603-23-1
Molecular Formula C20H30B2O4
Formula Weight 356.08
MDL Number MFCD02683500
Beilstein 8855562
Synonym cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1965103

 
alfa aesar™
L1965103
1g Each for $113.28

AAL1965106

 
alfa aesar™
L1965106
5g Each for $403.30
Description & Specifications

Specifications

Formula Weight 356.08
Synonym cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
Chemical Name or Material (E)-alpha,beta-Styrenediboronic acid bis(pinacol) ester
Assay 98%
Molecular Formula C20H30B2O4
MDL Number MFCD02683500
CAS 173603-23-1
Beilstein 8855562
Melting Point 69° to 73°C
Quantity 1g