Please login to your online account to display your discounted pricing

Alfa Aesar™ (E)-alpha-(4-Chlorophenyl)cinnamonitrile, 99%

$79.72 - $272.73

Chemical Identifiers

Chemical Name or Material (E)-alpha-(4-Chlorophenyl)cinnamonitrile
CAS 16610-81-4
Molecular Formula C15H10ClN
Formula Weight 239.7
MDL Number MFCD00082873
Beilstein 2331785
Synonym 2-(4-Chlorophenyl)-3-phenylacrylonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2564406

  SDS
alfa aesar™
B2564406
5g Each for $79.72

AAB2564414

  SDS
alfa aesar™
B2564414
25g Each for $272.73
Description & Specifications

Specifications

Formula Weight 239.7
Melting Point 113° to 118°C
Synonym 2-(4-Chlorophenyl)-3-phenylacrylonitrile
MDL Number MFCD00082873
CAS 16610-81-4
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (E)-alpha-(4-Chlorophenyl)cinnamonitrile
Odor Odorless
Quantity 5g
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 2331785
Molecular Formula C15H10ClN
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.