Please login to your online account to display your discounted pricing

Alfa Aesar™ (E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester, 98%

$299.18 - $299.18

Chemical Identifiers

Chemical Name or Material (E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester
CAS 307531-75-5
Molecular Formula C17H32B2O4
Formula Weight 322.06g/mol
MDL Number MFCD03093909
Synonym 1-cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxoboralan-2-yl)pentene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1964806

 
alfa aesar™
L19648-06
5g Each for $299.18
Description & Specifications

Specifications

Quantity 5g
Boiling Point 301° to 303°C
Quantity 5g
Flash Point 76°C (168°F)
Chemical Name or Material (E)-1-Pentene-1,2-diboronic acid bis(pinacol) ester
CAS 307531-75-5
Assay 98%
Molecular Formula C17H32B2O4
Formula Weight 322.06g/mol
MDL Number MFCD03093909
Synonym 1-cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxoboralan-2-yl)pentene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.