Please login to your online account to display your discounted pricing

Alfa Aesar™ (E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester, 98%

$103.76 - $406.36

Chemical Identifiers

Chemical Name or Material (E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester
CAS 185427-48-9
Molecular Formula C18H34B2O4
Formula Weight 336.09
MDL Number MFCD03093898
Beilstein 7768251
Synonym 1-cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hexene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1965003

 
alfa aesar™
L1965003
1g Each for $103.76

AAL1965006

 
alfa aesar™
L1965006
5g Each for $406.36
Description & Specifications

Specifications

Formula Weight 336.09
CAS 185427-48-9
Assay 98%
Molecular Formula C18H34B2O4
MDL Number MFCD03093898
Beilstein 7768251
Synonym 1-cis-1,2-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hexene
Boiling Point 102° to 104°C (0.08mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Chemical Name or Material (E)-1-Hexene-1,2-diboronic acid bis(pinacol) ester