missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Dystrophin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, affinity purification and electron microscopy applications.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS486P1ABX

 View more versions of this product

Catalog No. MS486P1ABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, affinity purification and electron microscopy applications with Thermo Scientific™ Dystrophin Ab-1, Mouse Monoclonal Antibody.

Dystrophin is a member of the spectrin/α-actinin family of actin-binding, triple helix rod-containing proteins. It is absent or greatly reduced in individuals with the X-linked recessive Duchenne's muscular dystrophy disorder, as well as in mice with the mdx (murine muscular dystrophy) mutation.

Host Species: Mouse

Clone: 1808

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Chicken, Xenopus and Torpedo. Others not known.

Epitope: Not determined

Immunogen: Acetylcholine receptor (AChR)-enriched membranes and peripheral membrane proteins from Torpedo nobiliana electric organ

Molecular Weight: 427 kDa

Positive Control: Skeletal muscle

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Affinity Purification
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Dystrophin Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG1
RUO
Primary
Chicken,Human,Murine,Rat,Xenopus
Immunofluorescence,Immunohistochemistry (Paraffin)
1808
Unlabeled
Concentrated
Acetylcholine receptor (AChR)-enriched membranes and peripheral membrane proteins from Torpedo nobiliana electric organ
100μL
Muscle Biology
Monoclonal