missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium sputtering target, 50.8mm dia x 3.18mm thick

$2,338.16 - $2,338.16

Chemical Identifiers

CAS 7429-91-6
MDL Number MFCD00010982
Chemical Name or Material Dysprosium sputtering target, 50.8mm dia. x 3.18mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45735KS SDS Alfa Aesar™
45735KS
1 ea. Each for $2,338.16
Specifications Description & Specifications

Chemical Identifiers

7429-91-6
MFCD00010982
Dysprosium sputtering target, 50.8mm dia. x 3.18mm thick

Specifications

1 ea.
Odorless
Dysprosium sputtering target, 50.8mm dia. x 3.18mm thick
7429-91-6
MFCD00010982

Safety and Handling

TSCA : TSCA