Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta)

Chemical Identifiers

Chemical Name or Material Dysprosium rod
CAS 7429-91-6
Name Note 12.7mm (0.5 in.) dia.
MDL Number MFCD00010982
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40286DH

  SDS
alfa aesar™
40286DH
25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40286DJ

  SDS
alfa aesar™
40286DJ
50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40286DM

  SDS
alfa aesar™
40286DM
100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40286HF

  SDS
alfa aesar™
40286HF
10mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Cast Rod, dia. is approximate
Quantity 25mm
Packaging Mylar under Argon
Chemical Name or Material Dysprosium rod
Odor Odorless
TSCA TSCA
Name Note 12.7mm (0.5 in.) dia.
Assay 99.9%
Assay Percent Notes (metals basis excluding Ta)
MDL Number MFCD00010982
CAS 7429-91-6
Storage Note 1
  • Packaged under argon
Weight ≈1.1g/mm

  • Packaged under argon

Safety and Handling

  • Packaged under argon