missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium powder, -200 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4591806

 View more versions of this product

Catalog No. AA4591806



Description

Description

  • Packaged under Argon
Specifications

Specifications

Dysprosium powder
Air & Moisture Sensitive
5g
Odorless
(REO)
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
Powder
−200 Mesh
Silver
7429-91-6
99.9%
Dy
UN3089
[Dy]
162.5
CHEBI:33377

Chemical Identifiers

7429-91-6
162.5
KBQHZAAAGSGFKK-UHFFFAOYSA-N
CHEBI:33377
[Dy]
Dy
MFCD00010982
23912
dysprosium
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378b-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents