missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium pieces, distilled dendritic, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1225706

 View more versions of this product

Catalog No. AA1225706



Description

Description

  • Packaged under argon
Specifications

Specifications

Dysprosium pieces
5g
7429-91-6
(REO)
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
[Dy]
162.5
CHEBI:33377
99.9%
Silver
Distilled dendritic
Odorless
Dy
UN3178
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
Pieces

Chemical Identifiers

7429-91-6
162.5
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
CHEBI:33377
[Dy]
Safety and Handling

Safety and Handling

GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents