Learn More
Dysprosium pieces, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 00010609
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Silver to Gray | |
7429-91-6 | |
Dy | |
UN3178 | |
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
[Dy] | |
162.50 | |
CHEBI:33377 | |
Odorless |
Dysprosium pieces | |
(REO) | |
MFCD00010982 | |
10 g | |
KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
dysprosium | |
23912 | |
99.9% | |
Pieces |
Chemical Identifiers
7429-91-6 | |
162.50 | |
KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
23912 | |
dysprosium |
Dy | |
MFCD00010982 | |
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
CHEBI:33377 | |
[Dy] |
Safety and Handling
GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.
GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber : 231-073-9
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only