Please login to your online account to display your discounted pricing

Dysprosium(III) trifluoromethanesulfonate, 99%, ACROS Organics™


Quantity 1g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC386120010

 View more versions of this product

Catalog No. AC386120010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex Dy)
Formula Weight 609.71
Physical Form Powder
Quantity 1g
Color White to gray
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00209583
CAS 139177-62-1
Assay Percent Range 99%
Chemical Name or Material Dysprosium(III) trifluoromethanesulfonate
Packaging Plastic bottle
Molecular Formula C3DyF9O9S3
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Dysprosium(III) trifluoromethanesulfonate
CAS 139177-62-1
Assay ≥98.5% (ex Dy)
Assay Percent Range 99%
Molecular Formula C3DyF9O9S3
Formula Weight 609.71
MDL Number MFCD00209583
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents