missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) oxide, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1131822

 View more versions of this product

Catalog No. AA1131822



Specifications

Specifications

Dysprosium(III) oxide
(REO)
14,3482
Powder
Reacton™
Hygroscopic
100g
1308-87-8
MFCD00010986
373
99.99%
7.81g/mL at 27°C
2340°C

Chemical Identifiers

1308-87-8
MFCD00010986
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents