missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dysprosium(III) oxide, 99.9% (REO) , Alfa Aesar™, REacton™

For Use in Cermets Used As Nuclear Reaction Control Rods

Manufacturer:  Thermo Fisher Scientific 1131909

 View more versions of this product

Catalog No. AA1131909



Specifications

Specifications

Dysprosium(III) oxide
99.9% (REO)
MFCD00010986
373
White
1308-87-8
Dy2O3
White powder or lump
Powder or Lump
10g

Chemical Identifiers

1308-87-8
Dy2O3
MFCD00010986
Safety and Handling

Safety and Handling

EINECSNumber : 215-164-0

Documents