Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) nitrate pentahydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Dysprosium(III) nitrate pentahydrate
CAS 10031-49-9
Purity Grade Reacton™
Formula Weight 438.59 (348.51 Anhydrous)
MDL Number MFCD00149683
UN Number UN1477
Merck Index 14,3482
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1131514

  SDS
alfa aesar™
1131514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1131522

  SDS
alfa aesar™
1131522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 438.59 (348.51 Anhydrous)
Melting Point 88.6°C
Odor Odorless
Merck Index 14,3482
UN Number UN1477
Chemical Name or Material Dysprosium(III) nitrate pentahydrate
TSCA TSCA
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Purity Grade Reacton™
pH 3
Assay 99.99%
Color Cream to pale yellow
Health Hazard 1 Danger
CAS 10031-49-9
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Assay Percent Notes (REO)
Physical Form Crystalline Aggregates
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Quantity 25g
MDL Number MFCD00149683
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.