Please login to your online account to display your discounted pricing

Dysprosium(III) nitrate pentahydrate, Light yellow crystalline aggregrates, 99.9% (REO), Alfa Aesar™

$59.96 - $59.96

Chemical Identifiers

Chemical Name or Material Dysprosium(III) nitrate pentahydrate
Name Note Rare Earth Oxides Content
CAS 10031-49-9
Physical Form Crystalline Aggregates
Assay Percent Range 99.9% (REO)
Molecular Formula Dy(NO3)3·5H2O
Formula Weight 438.59 (348.51 Anhydrous)
MDL Number MFCD00149683
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1292222

  SDS
alfa aesar
1292222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1292214

  SDS
alfa aesar
1292214
25g Each for $59.96
Description & Specifications

Specifications

Physical Form Crystalline Aggregates
Formula Weight 438.59 (348.51 Anhydrous)
Quantity 100g
Color Light yellow
DOT Information DOT Class: 5.1, Packing Group: III
Anhydrous or Hydrate Pentahydrate
EINECS Number 233-410-5
Name Note Rare Earth Oxides Content
Chemical Name or Material Dysprosium(III) nitrate pentahydrate
CAS 10031-49-9
Health Hazard 2 Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Assay Percent Range 99.9% (REO)
Molecular Formula Dy(NO3)3·5H2O
MDL Number MFCD00149683
Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.