Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) chloride, ultra dry, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Dysprosium(III) chloride
CAS 10025-74-8
Name Note Ultra dry
Formula Weight 268.86g/mol
MDL Number MFCD00010984
Merck Index 14,3482
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4711406

 
alfa aesar™
47114-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 3.67g/mL
Mesh Size −10 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Melting Point 718°C
Quantity 5g
Chemical Name or Material Dysprosium(III) chloride
Name Note Ultra dry
CAS 10025-74-8
Physical Form Beads
Assay 99.99%
Assay Percent Notes (REO)
Formula Weight 268.86g/mol
MDL Number MFCD00010984
Merck Index 14,3482
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Storage Note 1
  • Ampouled under argon
TSCA TSCA

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.

  • Ampouled under argon