missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) chloride hydrate, 99.99% (REO)

$46.95 - $161.31

Chemical Identifiers

MDL Number MFCD00010984
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1131706 View Documents Alfa Aesar™
1131706
5g Each for $46.95
 
AA1131714 View Documents Alfa Aesar™
1131714
25g Each for $161.31
 
AA1131722 View Documents Alfa Aesar™
1131722
100g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

MFCD00010984

Specifications

MFCD00010984
Wet Crystalline Aggregates
Hygroscopic
Dysprosium(III) chloride hydrate
268.86 (Anhydrous)
99.99%
5g
(REO)
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-039-9

TSCA : Yes

Recommended Storage : Ambient temperatures