missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) chloride hydrate, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1131714

 View more versions of this product

Catalog No. AA1131714



Specifications

Specifications

Dysprosium(III) chloride hydrate
MFCD00010984
Wet Crystalline Aggregates
Hygroscopic
(REO)
268.86 (Anhydrous)
99.99%
25g

Chemical Identifiers

MFCD00010984
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 233-039-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents