Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) chloride hydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Dysprosium(III) chloride hydrate
Formula Weight 268.86 (Anhydrous)
MDL Number MFCD00010984
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1131614

  SDS
alfa aesar™
1131614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1131622

  SDS
alfa aesar™
1131622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 268.86 (Anhydrous)
MDL Number MFCD00010984
Chemical Name or Material Dysprosium(III) chloride hydrate
Assay Percent Notes (REO)
Quantity 25g
TSCA TSCA
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Assay 99.9%
Sensitivity Hygroscopic
Physical Form Crystalline Aggregates
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.