missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) carbonate tetrahydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1528609

 View more versions of this product

Catalog No. AA1528609



Specifications

Specifications

Dysprosium(III) carbonate tetrahydrate
(REO)
MFCD00150229
BQRZSEZQNZPPPU-UHFFFAOYSA-H
dysprosium(3+);tricarbonate;tetrahydrate
25021600
Powder
Hygroscopic
38245-35-1
C3H8Dy2O13
dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]
577.084
577.06 (505.03 Anhydrous)
99.9%
10g

Chemical Identifiers

38245-35-1
577.084
BQRZSEZQNZPPPU-UHFFFAOYSA-H
25021600
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.O.O.O.[Dy+3].[Dy+3]
C3H8Dy2O13
MFCD00150229
dysprosium carbonate tetrahydrate, acmc-20al3d, dysprosium carbonate, dysprosium iii carbonate tetrahydrate, dysprosium carbonate-water 2/3/4, didysprosium 3+ ion tetrahydrate trico3
dysprosium(3+);tricarbonate;tetrahydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents