Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) carbonate tetrahydrate, 99.9% (REO)

$53.42 - $53.42

Chemical Identifiers

Chemical Name or Material Dysprosium(III) carbonate tetrahydrate
CAS 38245-35-1
Formula Weight 577.06 (505.03 Anhydrous)
MDL Number MFCD00150229
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1528609

 
alfa aesar™
1528609
10g Each for $53.42

AA1528618

 
alfa aesar™
1528618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1528630

 
alfa aesar™
1528630
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 577.06 (505.03 Anhydrous)
Chemical Name or Material Dysprosium(III) carbonate tetrahydrate
TSCA TSCA
Physical Form Powder
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00150229
CAS 38245-35-1
Quantity 10g