Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) bromide, ultra dry, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Dysprosium(III) bromide
CAS 14456-48-5
Name Note Ultra dry
Formula Weight 402.23
MDL Number MFCD00016069
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4712703

  SDS
alfa aesar™
4712703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4712706

  SDS
alfa aesar™
4712706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 402.23
Packaging Ampule
Assay 99.99%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Notes (metals basis)
Sensitivity Hygroscopic
Melting Point 881°C
Mesh Size −10 Mesh
Chemical Name or Material Dysprosium(III) bromide
Physical Form Beads
TSCA TSCA
Health Hazard 1 Warning
Storage Note 1
  • Ampouled under argon
Name Note Ultra dry
MDL Number MFCD00016069
CAS 14456-48-5
Boiling Point 1480°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon