Please login to your online account to display your discounted pricing

Alfa Aesar™ Dysprosium(III) acetate tetrahydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Dysprosium(III) acetate tetrahydrate
CAS 15280-55-4
Purity Grade Reacton™
Formula Weight 411.68 (339.62 Anhydrous)
MDL Number MFCD00013033
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1459522

  SDS
alfa aesar™
1459522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1459514

  SDS
alfa aesar™
1459514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 411.68 (339.62 Anhydrous)
Quantity 100g
Odor Odorless
CAS 15280-55-4
Chemical Name or Material Dysprosium(III) acetate tetrahydrate
MDL Number MFCD00013033
Physical Form Crystalline Aggregates
Melting Point 120°C (dec.)
Purity Grade Reacton™
TSCA TSCA
Assay 99.9%
Assay Percent Notes (REO)