missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dysprosium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01321914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dysprosium(III) 2,4-pentanedionate hydrate | |
Powder | |
(REO) | |
MFCD00013489 | |
VABPDCUJKLETFM-UHFFFAOYSA-N | |
dysprosium;(Z)-4-hydroxypent-3-en-2-one | |
121233142 | |
99.9% | |
Odorless |
14637-88-8 | |
25 g | |
C15H21DyO6 | |
dysprosium acetylacetonate hydrate, dysprosium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
[Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
459.83 | |
459.83 (Anhydrous) | |
Reacton™ | |
Hygroscopic |
Chemical Identifiers
14637-88-8 | |
459.83 | |
VABPDCUJKLETFM-UHFFFAOYSA-N | |
121233142 | |
[Dy+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21DyO6 | |
MFCD00013489 | |
dysprosium acetylacetonate hydrate, dysprosium,tris 2,4-pentanedionato-ko2,ko4-, oc-6-11 | |
dysprosium;(Z)-4-hydroxypent-3-en-2-one |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only